Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR39 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | GPR39 |
---|---|
Concentration | 0.32 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-4048
![]() ![]() |
Proteintech Group Inc
233261AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The G-protein-coupled receptor (GPCR) superfamily is comprised of an estimated 600-1,000 members and is the largest known class of molecular targets with proven therapeutic value. G protein-coupled receptor 39 is an orphan receptor in the Ghrelin family pathway. Various studies have shown that GPR39 plays important roles in hypertension, adipogenesis and that overexpression of GPR39 contributes to maligt development of human esophageal squamous cell carcinoma.Specifications
GPR39 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
O43194 | |
2863 | |
GPR39 Fusion Protein Ag19913 | |
Primary | |
-20°C | |
GPR39 |
0.32 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
G protein coupled receptor 39, GPR39 | |
GPR39 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title