Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP118996
Description
GPR4 Polyclonal specifically detects GPR4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
GPR4 | |
Polyclonal | |
Unconjugated | |
Antiserum with No Preservative | |
GPR4 | |
Synthetic peptide derived from GPR4 protein | |
0.2 mL | |
GPCR, Signal Transduction | |
2828 | |
Reconstitute with 0.2 ml distilled water. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
LYOPH | |
Western Blot 1:200-1:1000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 | |
G protein-coupled receptor 4, GPR19, G-protein coupled receptor 19, G-protein coupled receptor 4 | |
Rabbit | |
Unpurified | |
RUO | |
Primary | |
Reacts specifically with human 41 kDa protein | |
Human, Mouse, Rat | |
Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction