Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR40 (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA577398
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
This gene encodes a member of the GP40 family of G protein-coupled receptors that are clustered together on chromosome 19. The encoded protein is a receptor for medium and long chain free fatty acids and may be involved in the metabolic regulation of insulin secretion. Polymorphisms in this gene may be associated with type 2 diabetes.
Specifications
GPR40 (extracellular) | |
Polyclonal | |
Unconjugated | |
Ffar1 | |
GPR40,FFAR1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
266607 | |
-20°C | |
Lyophilized |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
Q8K3T4 | |
Ffar1 | |
Peptide (C)NVASFINPDLEGSWRK corresponding to amino acid residues 244-259 of rat free fatty acid receptor 1 | |
50 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction