Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPR40 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518381
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
This gene encodes a member of the GP40 family of G protein-coupled receptors that are clustered together on chromosome 19. The encoded protein is a receptor for medium and long chain free fatty acids and may be involved in the metabolic regulation of insulin secretion. Polymorphisms in this gene may be associated with type 2 diabetes.
Specifications
GPR40 | |
Polyclonal | |
Unconjugated | |
FFAR1 | |
FFA1R; FFAR; Ffar1; Free fatty acid receptor 1; G protein-coupled receptor 40; G protein-coupled receptor GPR40; GPCR40; GPR40; G-protein coupled receptor 40; HGNC:4498 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
2864 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O14842 | |
FFAR1 | |
Synthetic peptide sequence (CAARTQGGKSQK) corresponding to the C-terminus amino acids of FFAR1 (aa 289-300). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction