Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR41 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560791
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560791 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560791 Supplier Invitrogen™ Supplier No. PA560791
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DFHELLRRLC GLWGQWQQES SMELKEQKGG EEQRADRPAE RKTSEHSQGC GTGGQVAC Highest antigen sequence identity to the following orthologs: Mouse - 32%, Rat - 33%.

G protein-coupled receptors (GPRs), also known as seven transmembrane receptors, heptahelical receptors or 7TM receptors, comprise a superfamily of proteins that play a role in many different stimulus-response pathways. GPRs translate extracellular signals into intracellular signals (a process called G-protein activation) and they respond to a variety of signaling molecules, such as hormones and neurotransmitters. GPR41 (G-protein coupled receptor 41), also known as FFAR3 (Free fatty acid receptor 3), is a 346 amino acid multi-pass membrane protein that belongs to the G protein-coupled receptor family. Expressed at high levels in adipose tissue and at lower levels throughout the body, GPR41 functions as a receptor for short chain fatty acids via elevation of intracellular calcium levels and inhibition of adenylyl cyclase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR41
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FFAR3
Gene Accession No. O14843
Gene Alias FFA3R; FFAR3; Free fatty acid receptor 3; G protein-coupled receptor 41; Gm478; Gpcr41; GPR41; GPR42; G-protein coupled receptor 41; putative G protein-coupled receptor GPR41
Gene Symbols FFAR3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GPR41. Recombinant protein control fragment (Product #RP-100019).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2865
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.