Learn More
GPR43 (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5111779
Description
Reconstitute antibody with 50 μL of double distilled water (DDW). The reconstituted solution can be stored at 4 degrees C for up to 1 week. For longer periods, small aliquots should be stored at -20 degrees C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min). Peptide sequence is: (C)TEQVRSG NEIT(S)YEN Homology: Human only.
G protein-coupled receptors (GPCRs, or GPRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins. GPCR GPR43 is a member of this family (subfamily carboxylic acid). It expression has been reported primarily in immune cells including monocytes and neutrophils. An EST has been isolated from a human pancreatic library.
Specifications
GPR43 (extracellular) | |
Polyclonal | |
Unconjugated | |
FFAR2 | |
FFA2; FFA2R; Ffar2; Free fatty acid receptor 2; GPCR43; Gpr43; G-protein coupled receptor 43; Leukocyte-specific STAT-induced GPCR; Lssig | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2867 | |
-20°C | |
Lyophilized |
Flow Cytometry, Western Blot | |
0.8 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
O15552 | |
FFAR2 | |
Peptide corresponding to amino acid residues 153-167 of human FFAR2 (2 nd extracellular loop) | |
50 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.