Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR43 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100944
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100944 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100944 Supplier Invitrogen™ Supplier No. PA5100944
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GPR43.

G protein-coupled receptors (GPCRs, or GPRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins. GPCR GPR43 is a member of this family (subfamily carboxylic acid). It expression has been reported primarily in immune cells including monocytes and neutrophils. An EST has been isolated from a human pancreatic library.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR43
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FFAR2
Gene Accession No. O15552
Gene Alias fatty acid receptor 2; FFA2; FFA2R; FFAR2; free fatty acid activated receptor 2; free fatty acid receptor 2; G protein-coupled receptor 43; GPCR43; Gpr43; G-protein coupled receptor 43; leukocyte-specific STAT-induced GPCR; Lssig
Gene Symbols FFAR2
Host Species Rabbit
Immunogen A synthesized peptide derived from human FFAR2(Accession O15552), corresponding to amino acid residues V290-E330.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2867
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.