Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR43 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982954
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2954 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2954 Supplier Bioss Supplier No. BS13536R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

G protein-coupled receptors (GPCRs, or GPRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins. GPCR GPR43 is a member of this family (subfamily carboxylic acid). It expression has been reported primarily in immune cells including monocytes and neutrophils. An EST has been isolated from a human pancreatic library.

Specifications

Antigen GPR43
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene FFAR2
Gene Accession No. O15552
Gene Alias fatty acid receptor 2; FFA2; FFA2R; Ffar2; free fatty acid activated receptor 2; Free fatty acid receptor 2; G protein-coupled receptor 43; GPCR43; GPR43; G-protein coupled receptor 43; Leukocyte-specific STAT-induced GPCR; Lssig
Gene Symbols FFAR2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GPR43.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2867
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.