Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR45 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533723
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533723 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533723 Supplier Invitrogen™ Supplier No. PA533723
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gibbon, Monkey, Marmoset, Rabbit (100%) Gorilla, Bat, Lizard (94%).

This intronless gene encodes a member of the G protein-coupled receptor (GPCR) family. Members of this protein family contain seven putative transmembrane domains and may mediate signaling processes to the interior of the cell via activation of heterotrimeric G proteins. This protein may function in the central nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR45
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene GPR45
Gene Accession No. Q9Y5Y3
Gene Alias 9230112G11Rik; AW046772; G protein-coupled receptor 45; GPR45; high-affinity lysophosphatidic acid receptor; probable G-protein coupled receptor 45; PSP24; PSP24(ALPHA); PSP24-1; PSP24A; PSP24alpha; PSP24-alpha
Gene Symbols GPR45
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from 2nd extracellular domain of human GPR45.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100342689, 11250
Target Species Human, Rabbit, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.