Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR45 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118811
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118811 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5118811 Supplier Invitrogen™ Supplier No. PA5118811
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GPR45.

This intronless gene encodes a member of the G protein-coupled receptor (GPCR) family. Members of this protein family contain seven putative transmembrane domains and may mediate signaling processes to the interior of the cell via activation of heterotrimeric G proteins. This protein may function in the central nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR45
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GPR45
Gene Accession No. Q9EQQ4, Q9Y5Y3
Gene Alias 9230112G11Rik; AW046772; G protein-coupled receptor 45; GPR45; high-affinity lysophosphatidic acid receptor; probable G-protein coupled receptor 45; PSP24; PSP24(ALPHA); PSP24-1; PSP24A; PSP24alpha; PSP24-alpha
Gene Symbols GPR45
Host Species Rabbit
Immunogen A synthesized peptide derived from human GPR45(Accession Q9Y5Y3), corresponding to amino acid residues F117-L167.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11250, 93690
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.