Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR45 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | GPR45 |
|---|---|
| Dilution | Western Blot 1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
GPR45 Polyclonal specifically detects GPR45 in Human samples. It is validated for Western Blot.Specifications
| GPR45 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| GPCR | |
| PBS buffer, 2% sucrose | |
| 11250 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1.0 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| G protein-coupled receptor 45, high-affinity lysophosphatidic acid receptor, probable G-protein coupled receptor 45, PSP24, PSP24(ALPHA), PSP24-1, PSP24A, PSP24-alpha | |
| The immunogen is a synthetic peptide directed towards the N-terminal region of Human GPR45 (NP_009158). Peptide sequence LNTSNASDSGSTQLPAPLRISLAIVMLLMTVVGFLGNTVVCIIVYQRPAM | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title