Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR49/LGR5 Polyclonal Antibody, Biotin, Bioss
SDP

Catalog No. 502279623
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-9623 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-9623 Supplier Bioss Supplier No. BS20747RBIOTIN
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GPR49/LGR5 Polyclonal antibody specifically detects GPR49/LGR5 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen GPR49/LGR5
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Biotin
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene Lgr5
Gene Accession No. D4AC13, O75473, Q9Z1P4
Gene Alias Fex; follicle expressed hormone; G protein-coupled receptor 49; GPR49; GPR67; G-protein coupled receptor 49; G-protein coupled receptor 67; G-protein coupled receptor HG38; GRP49; HG38; leucine rich repeat containing G protein coupled receptor 5; leucine rich repeat containing G protein-coupled receptor 5; leucine-rich repeat containing G protein-coupled receptor 5; leucine-rich repeat-containing G protein-coupled receptor 5; leucine-rich repeat-containing G-protein coupled receptor 5; Lgr5; orphan G protein-coupled receptor HG38; orphan G-protein coupled receptor FEX
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GPR49/LGR5
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14160, 299802, 8549
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.