Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR50 Antibody (461129), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4645W
Description
GPR50 Monoclonal antibody specifically detects GPR50 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOFSpecifications
GPR50 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
461129 | |
Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
G protein-coupled receptor 50H9Mel1c, melatonin-related receptor, MGC125342 | |
NS0 mouse myeloma cell line transfected with human GPR50, Met1-Val617, Accession # Q13585 | |
0.1 mL | |
GPCR, Neuroscience | |
9248 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction