Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR50 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | GPR50 |
---|---|
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Regulatory Status | RUO |
Description
GPR50 Polyclonal specifically detects GPR50 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
GPR50 | |
Unconjugated | |
RUO | |
Human | |
G protein-coupled receptor 50H9Mel1c, melatonin-related receptor, MGC125342 | |
GPR50 | |
IgG | |
Affinity Purified |
Polyclonal | |
Rabbit | |
GPCR, Neuroscience | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
9248 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:PTKPAASQLESDTIADLPDPTVVTTSTNDYHDVVVIDVEDDPDEMAV | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title