Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR50 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111396
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111396 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111396 Supplier Invitrogen™ Supplier No. PA5111396
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PTKPAASQLE SDTIADLPDP TVVTTSTNDY HDVVVIDVED DPDEMAV Highest antigen sequence indentity to the following orthologs: Rat - 37%, Mouse - 34%.

Melatonin-related receptor expression has been documented from human CNS tissues, specifically in hypothalamus and pituitary, but not in human peripheral tissues. In animals, expression has also been seen in the CNS, and peripheral tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR50
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GPR50
Gene Accession No. Q13585
Gene Alias G protein-coupled receptor 50; Gpr50; G-protein-coupled receptor 50; H9; Mel1c; melatonin-related receptor; MTNRL
Gene Symbols GPR50
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human GPR50. Recombinant protein control fragment (Product #RP-106565).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9248
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.