Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR50 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533732
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533732 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533732 Supplier Invitrogen™ Supplier No. PA533732
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey, Marmoset, Sheep, Dog (100%) Gibbon, Mouse, Rat, Panda, Bovine, Rabbit (94%) Bat (88%).

Melatonin-related receptor expression has been documented from human CNS tissues, specifically in hypothalamus and pituitary, but not in human peripheral tissues. In animals, expression has also been seen in the CNS, and peripheral tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR50
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene GPR50
Gene Accession No. Q13585, Q28558
Gene Alias G protein-coupled receptor 50; Gpr50; G-protein-coupled receptor 50; H9; Mel1c; melatonin-related receptor; MTNRL
Gene Symbols GPR50
Host Species Rabbit
Immunogen Synthetic 17 amino acid peptide from 3rd cytoplasmic domain of human GPR50.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 443023, 492213, 9248
Target Species Human, Canine, Monkey, Sheep
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.