Learn More
GPR50 Polyclonal Antibody, Invitrogen™
Description
PA1-32876 detects GPR50 from human samples.
Specifications
Specifications
| Antigen | GPR50 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene | GPR50 |
| Gene Accession No. | Q13585 |
| Gene Alias | G protein-coupled receptor 50; Gpr50; G-protein-coupled receptor 50; H9; Mel1c; melatonin-related receptor; MTNRL |
| Gene Symbols | GPR50 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.