Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR52 Antibody (582671), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB10357MFV450
Description
GPR52 Monoclonal antibody specifically detects GPR52 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
GPR52 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
0.1 mL | |
GPCR | |
9293 | |
Store at 4C in the dark. |
Flow Cytometry, CyTOF | |
582671 | |
Flow Cytometry, CyTOF-ready | |
G protein-coupled receptor 52, MGC111751, probable G-protein coupled receptor 52 | |
NS0 mouse myeloma cell line transfected with human GPR52, Met1-Ile361, Accession # Q9Y2T5 | |
RUO | |
Primary | |
Human | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction