Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR52 Polyclonal Antibody

Catalog No. PIPA570593
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570593 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570593 Supplier Invitrogen™ Supplier No. PA570593

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 82%; Guinea Pig: 93%; Human: 100%; Mouse: 79%; Pig: 100%; Rat: 90%.

GPR52 is a member of the G protein-coupled receptor (GPR) family play important roles in signal transduction from the external environment to the inside of the cell.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR52
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GPR52
Gene Accession No. 0
Gene Alias Axor10; EG620246; G protein-coupled receptor 52; Gpr52; G-protein coupled receptor 52; MGC111751; probable G-protein coupled receptor 52
Gene Symbols GPR52
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of rat GPR52.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 684623
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.