Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR55 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP289010
Description
GPR55 Polyclonal specifically detects GPR55 in Mouse, Rat samples. It is validated for Western Blot.Specifications
GPR55 | |
Polyclonal | |
PBS containing 50% glycerol | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Mouse, Rat | |
Purified |
Western Blot | |
Unconjugated | |
G protein-coupled receptor 55, G-protein coupled receptor 55 | |
Synthetic peptide corresponding to the sequence near the N-terminus of rat GPR55. | |
50 μL | |
GPCR | |
9290 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction