Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR56 Antibody (2446B), PE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB46361P
Description
GPR56 Monoclonal antibody specifically detects GPR56 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
GPR56 | |
Monoclonal | |
PE | |
PBS | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
2446B | |
Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
BFPP, G protein-coupled receptor 56,7-transmembrane protein with no EGF-like N-terminal domains-1, Protein TM7XN1, TM7LN4G-protein coupled receptor 56, TM7XN1DKFZp781L1398 | |
Chinese hamster ovary cell line CHO-derived recombinant human GPR56, Met1-Val345, Accession # Q9Y653 | |
0.1 mL | |
GPCR, Growth and Development, Neuronal Cell Markers | |
9289 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction