Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR56 Antibody (776123), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4636I
Description
GPR56 Monoclonal antibody specifically detects GPR56 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
GPR56 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
776123 | |
Flow Cytometry, CyTOF-ready | |
BFPP, G protein-coupled receptor 56,7-transmembrane protein with no EGF-like N-terminal domains-1, Protein TM7XN1, TM7LN4G-protein coupled receptor 56, TM7XN1DKFZp781L1398 | |
Chinese hamster ovary cell line CHO-derived recombinant human GPR56, Met1-Val342, Accession # Q9Y653 | |
0.1 mL | |
GPCR, Growth and Development, Neuronal Cell Markers | |
9289 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction