Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR56 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533739
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533739 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533739 Supplier Invitrogen™ Supplier No. PA533739
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human (100%) Chimpanzee, Orangutan, Monkey (94%) Gorilla, Marmoset (88%) Elephant (81%).

GPR56 is a member of the G protein-coupled receptor family. The protein contains 7 transmembrane domains and a mucin-like domain in the N-terminal region. The gene is implicated in the regulation of brain cortical patterning. The protein binds specifically to transglutaminase 2 in the extracellular space. Expression of this gene is downregulated in melanoma cell lines, and overexpression of this gene can suppress tumor growth and metastasis. Mutations in this gene result in bilateral frontoparietal polymicrogyria. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR56
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene ADGRG1
Gene Accession No. Q9Y653
Gene Alias 7-transmembrane protein with no EGF-like N-terminal domains-1; Adgrg1; ADGRG1 CT; ADGRG1 C-terminal fragment; ADGRG1 NT; ADGRG1 N-terminal fragment; ADGRG1-CT; adhesion G protein-coupled receptor G1; adhesion G-protein coupled receptor G1; BFPP; BPPR; Cyt28; DKFZp781L1398; G protein-coupled receptor 56; Gpr56; GPR56 7TM; GPR56 CT; GPR56 C-terminal fragment; GPR56 extracellular subunit; GPR56 NT; GPR56 N-terminal fragment; GPR56 seven-transmembrane subunit; GPR56 subunit alpha; GPR56 subunit beta; G-protein coupled receptor 56; Protein TM7XN1; secretin receptor; serpentine receptor cyt28; testicular tissue protein Li 77; TM7LN4; TM7XN1; UNQ540/PRO1083
Gene Symbols ADGRG1
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from N-terminal extracellular domain of human GPR56.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9289
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.