Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590013
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590013 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590013 Supplier Invitrogen™ Supplier No. PA590013
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WRHAHQIALQ QHCLAPPHLA ATRKGVGTLA VVLGTFGASW LPFAIYCVVG SHEDPAVYTY ATLLPATYNS MINPIIYAFR NQEIQRALWL LLCGCFQSKV PFRSRSPSEV; Positive Samples: U-251MG; Cellular Location: Cell membrane, Multi-pass membrane protein.

The G-protein-coupled receptor (GPCR) superfamily is comprised of an estimated 600-1,000 members and is the largest known class of molecular targets with proven therapeutic value. GPR6 is an orphan, constitutively active G-coupled protein receptor, involved in a wide array of functions such as neurite growth, amyloid processing and appetitive learning. GPR6 is predominantly expressed in neurons in the striatum of the brain. GPR6 was found to be expressed on the cell surface and in intracellular compartments.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR6
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene GPR6
Gene Accession No. P46095, Q6YNI2
Gene Alias AI852874; G protein-coupled receptor 6; Gm233; GPR6; G-protein coupled receptor 6; RCNL3; Sphingosine 1-phosphate receptor GPR6
Gene Symbols GPR6
Host Species Rabbit
Immunogen Recombinant protein (or fragment)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140741, 2830
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.