Learn More
Invitrogen™ GPR61 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA3038
Description
IHC(P) analysis shows positive staining of GPR61 in human pancreatic islets and enteric ganglion. WB analysis shows GPR61 in glyco-protein enriched fractions from mock transfected or GPR61 overexpressed 293 cells. The GPR61 antiserum showed specific detection of GPR61 over other tested GPRs. GPR61 exists in different glycosylated forms and can form dimers and oligomers. Consequently it appears as a long smear on WB or is detected at multiple molecular weights.
G protein-coupled receptors (GPCRs, or GPRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins. GPCR GPR61 is a member of this family (subfamily Orphan-A). It has been reported to be expressed in several regions of brain, including caudate, putamen, and thalamus.
Specifications
GPR61 | |
Polyclonal | |
Unconjugated | |
GPR61 | |
BALGR; biogenic amine receptor-like GPCR; Biogenic amine receptor-like G-protein coupled receptor; G protein-coupled receptor 61; GPCR3; GPR61; G-protein coupled receptor 61; probable G-protein coupled receptor 61 | |
Rabbit | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Conc. Not Determined | |
whole serum with 0.05% sodium azide | |
Q9BZJ8 | |
GPR61 | |
KLH conjugated COOH-terminal amino acids of 432-452 (SDIIMSDSYLRPAASPRLES). | |
RUO | |
83873 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.