Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR61 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA120456
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA120456 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA120456 Supplier Thermo Scientific Supplier No. PA120456

Rabbit Polyclonal Antibody

PA1-20456 detects G Protein-Coupled Receptor GPR61 in Human samples.

G protein-coupled receptors (GPCRs, or GPRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins. GPCR GPR61 is a member of this family (subfamily Orphan-A). It has been reported to be expressed in several regions of brain, including caudate, putamen, and thalamus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR61
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene GPR61
Gene Accession No. Q9BZJ8
Gene Alias BALGR; biogenic amine receptor-like GPCR; Biogenic amine receptor-like G-protein coupled receptor; G protein-coupled receptor 61; GPCR3; GPR61; G-protein coupled receptor 61; probable G-protein coupled receptor 61
Gene Symbols GPR61
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from N-terminal extracellular domain of human GPR61
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83873
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.