Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR62 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533745
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533745 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533745 Supplier Invitrogen™ Supplier No. PA533745
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human (100%) Marmoset (88%) Elephant, Horse (81%).

GPR62 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR62
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene GPR62
Gene Accession No. Q9BZJ7
Gene Alias G protein-coupled receptor 62; GPCR8; GPR62; G-protein coupled receptor 62; G-protein coupled receptor GPCR8; G-protein coupled receptor KPG_005; hGPCR8; KPG_005; MGC26943; probable G-protein coupled receptor 62
Gene Symbols GPR62
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from N-terminal extracellular domain of human GPR62.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 118442
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.