Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR62 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA3048
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA3048 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA3048 Supplier Thermo Scientific Supplier No. PA3048

Rabbit Polyclonal Antibody

IHC(P) analysis shows positive staining of GPR62 in human pancreatic islets. WB analysis shows GPR62 in glyco-protein enriched fractions from mock transfected or GPR62 overexpressed 293 cells. GPR62 exists in different glycosylated forms.

GPR62 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR62
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 0.05% sodium azide
Gene GPR62
Gene Accession No. Q9BZJ7
Gene Alias G protein-coupled receptor 62; GPCR8; GPR62; G-protein coupled receptor 62; G-protein coupled receptor GPCR8; G-protein coupled receptor KPG_005; hGPCR8; KPG_005; MGC26943; probable G-protein coupled receptor 62
Gene Symbols GPR62
Host Species Rabbit
Immunogen KLH conjugated COOH-terminal amino acids of 349-368 (PELAGGRSPAYQGPPESSLS)
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 118442
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.