Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR63 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533749
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533749 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533749 Supplier Invitrogen™ Supplier No. PA533749
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Marmoset, Mouse, Rat, Elephant, Panda, Bovine, Bat, Horse (100%).

G protein-coupled receptors (GPCRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins. GPR63 is a member of this family (subfamily: lysophospholipid/lysosphingolipid) and its expression has been reported in several brain regions, including caudate, frontal cortex, hypothalamus, midbrain, and thalamus, as well as in small intestine, stomach, and thymus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR63
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene GPR63
Gene Accession No. Q9BZJ6, Q9EQQ3
Gene Alias brain expressed G-protein-coupled receptor PSP24 beta; G protein-coupled receptor 63; G protein-coupled receptor PSP24-2; Gpr63; Probable G-protein coupled receptor 63; PSP24(beta); PSP24-2; PSP24B; PSP24beta; PSP24-beta
Gene Symbols GPR63
Host Species Rabbit
Immunogen Synthetic 13 amino acid peptide from 1st extracellular domain of human GPR63.
Purification Method Antigen Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100071441, 297952, 517696, 81006, 81491
Target Species Human, Mouse, Rat, Bovine, Horse, Bat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.