Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR63 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA571989
Description
G protein-coupled receptors (GPCRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins. GPR63 is a member of this family (subfamily: lysophospholipid/lysosphingolipid) and its expression has been reported in several brain regions, including caudate, frontal cortex, hypothalamus, midbrain, and thalamus, as well as in small intestine, stomach, and thymus.
Specifications
GPR63 | |
Polyclonal | |
Unconjugated | |
GPR63 | |
GPR63; PSP24(beta); PSP24-2; PSP24-beta; PSP24B; G protein-coupled receptor 63 | |
Rabbit | |
Protein A | |
RUO | |
81491 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1.09 mg/mL | |
PBS with 0.09% sodium azide | |
Q9BZJ6 | |
GPR63 | |
KLH conjugated synthetic peptide between 381-410 amino acids from the C-terminal region of human GPR63 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction