Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR64 Polyclonal Antibody

GPR64 Polyclonal Antibody
Supplier: Thermo Scientific PA533752
Description
GPR64 Polyclonal Antibody for IHC (P)
Specifications
| GPR64 | |
| Polyclonal | |
| Unconjugated | |
| ADGRG2 | |
| ADGRG2; adhesion G protein-coupled receptor G2; adhesion G-protein coupled receptor G2; AW212196; B830041D06Rik; CTD-2245E12.1; EDDM6; epididymal protein 6; epididymis-specific protein 6; FLJ00282; G protein-coupled receptor 64; G protein-coupled receptor, epididymis-specific (seven transmembrane family); Gpr64; G-protein coupled receptor 64; He6; Human epididymis-specific protein 6; LNB-TM7 heptahelical receptor Me6; Me6; MGC104454; MGC138738; MGC138739; Mouse epididymis-specific protein 6; Rat epididymis-specific protein 6; Re6; re6 receptor; TM7LN2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100157214, 10149, 491763 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q8IZP9 | |
| ADGRG2 | |
| Synthetic 16 amino acid peptide from C-terminus of human GPR64. | |
| 50 μg | |
| Primary | |
| Human, Canine, Pig, Bat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction