Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR64 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565594
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565594 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565594 Supplier Invitrogen™ Supplier No. PA565594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NASGVKPQRNI CNLSSICNDS AFFRGEIMFQ YDKESTVPQN QHITNGTLTG VLSLSELKRS ELNKTLQTLS ETYFIMCATA EAQSTLNCTF TIKLNNTMNA CAVIAALERV KIRPMEHCCC SVRIPCP Highest antigen sequence identity to the following orthologs - mouse 76%, rat 75%.

Could be involved in a signal transduction pathway controlling epididymal function and male fertility.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR64
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ADGRG2
Gene Accession No. Q8IZP9
Gene Alias ADGRG2; adhesion G protein-coupled receptor G2; adhesion G-protein coupled receptor G2; AW212196; B830041D06Rik; CTD-2245E12.1; EDDM6; epididymal protein 6; epididymis-specific protein 6; FLJ00282; G protein-coupled receptor 64; G protein-coupled receptor, epididymis-specific (seven transmembrane family); Gpr64; G-protein coupled receptor 64; He6; Human epididymis-specific protein 6; LNB-TM7 heptahelical receptor Me6; Me6; MGC104454; MGC138738; MGC138739; Mouse epididymis-specific protein 6; Rat epididymis-specific protein 6; Re6; re6 receptor; TM7LN2
Gene Symbols ADGRG2
Host Species Rabbit
Immunogen Recombinant Human GPR64. Recombinant protein control fragment (Product #RP-104889).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10149
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.