Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR64 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA132497
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA132497 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA132497 Supplier Invitrogen™ Supplier No. PA132497
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-32497 detects GPCR GPR64 from human samples.

Could be involved in a signal transduction pathway controlling epididymal function and male fertility.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR64
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene ADGRG2
Gene Accession No. Q8CJ11, Q8CJ12, Q8IZP9
Gene Alias ADGRG2; adhesion G protein-coupled receptor G2; adhesion G-protein coupled receptor G2; AW212196; B830041D06Rik; CTD-2245E12.1; EDDM6; epididymal protein 6; epididymis-specific protein 6; FLJ00282; G protein-coupled receptor 64; G protein-coupled receptor, epididymis-specific (seven transmembrane family); Gpr64; G-protein coupled receptor 64; He6; Human epididymis-specific protein 6; LNB-TM7 heptahelical receptor Me6; Me6; MGC104454; MGC138738; MGC138739; Mouse epididymis-specific protein 6; Rat epididymis-specific protein 6; Re6; re6 receptor; TM7LN2
Gene Symbols ADGRG2
Host Species Rabbit
Immunogen Synthetic 20 amino acid peptide from C-terminus of human GPR64.
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100754781, 10149, 237175, 266735
Target Species Human, Mouse, Rat, Monkey, Hamster
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.