Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR64 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982090
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2090 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2090 Supplier Bioss Supplier No. BS12268R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Could be involved in a signal transduction pathway controlling epididymal function and male fertility.

Specifications

Antigen GPR64
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ADGRG2
Gene Accession No. Q8IZP9
Gene Alias ADGRG2; adhesion G protein-coupled receptor G2; Adhesion G-protein coupled receptor G2; CTD-2245E12.1; EDDM6; epididymal protein 6; FLJ00282; G protein-coupled receptor 64; G protein-coupled receptor, epididymis-specific (seven transmembrane family); GPR64; G-protein coupled receptor 64; HE6; Human epididymis-specific protein 6; MGC104454; MGC138738; MGC138739; TM7LN2
Gene Symbols ADGRG2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human G protein coupled receptor 64.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10149
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.