Learn More
Invitrogen™ GPR68 Polyclonal Antibody

Description
Western blot should be visualized using a high sensitivity secondary/substrate system (i.e SuperSignal West Femto Maximum Sensitivity Substrate). A suggested positive control for this product is OGR1 transfected cell lysate.
Specifications
Specifications
| Antigen | GPR68 |
| Applications | Immunohistochemistry, Neutralization, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.08% sodium azide |
| Gene | GPR68 |
| Gene Accession No. | Q15743 |
| Gene Alias | BB131428; Brgr1; Brgrb; G protein-coupled receptor 68; GPR12A; Gpr68; G-protein coupled receptor 68; Ogr1; OGR-1; ovarian cancer G protein-coupled receptor, 1; ovarian cancer G-protein coupled receptor 1; Sphingosylphosphorylcholine receptor |
| Gene Symbols | GPR68 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.