Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR75 Polyclonal Antibody

GPR75 Polyclonal Antibody
Supplier: Thermo Scientific PA533762
Description
GPR75 Polyclonal Antibody for IHC (P)
Specifications
| GPR75 | |
| Polyclonal | |
| Unconjugated | |
| GPR75 | |
| G protein-coupled receptor 75; GPR75; GPRchr2; GPR-chr2; Probable G-protein coupled receptor 75; WI31133; WI-31133 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100066961, 100517153, 10936, 539470 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| O95800 | |
| GPR75 | |
| Synthetic 16 amino acid peptide from 2nd cytoplasmic domain of human GPR75. | |
| 50 μg | |
| Primary | |
| Human, Bovine, Pig, Horse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction