Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPR75 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533759
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey, Rat, Bovine, Bat, Hamster, Elephant, Panda, Horse, Rabbit, Pig, Opossum, Chicken, Lizard, Xenopus (100%).
GPR75 is an Orphan-A GPCR with an unknown ligand. G protein-coupled receptor GPR75 has been reported to be expressed in brain and eye. ESTs have been isolated from human brain, eye, nerve, skin, and stomach libraries.
Specifications
| GPR75 | |
| Polyclonal | |
| Unconjugated | |
| GPR75 | |
| G protein-coupled receptor 75; GPR75; GPRchr2; GPR-chr2; Probable G-protein coupled receptor 75; WI31133; WI-31133 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100066961, 100347723, 100517153, 100766988, 10936, 428544, 498434, 539470 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| O95800 | |
| GPR75 | |
| Synthetic 19 amino acid peptide from C-terminus of human GPR75. | |
| 50 μg | |
| Primary | |
| Human, Rat, Rabbit, Monkey, Bovine, Hamster, Pig, Horse, Chicken, Frog, Bat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction