Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPR78 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533764
Description
Percent identity with other species by BLAST analysis: Human, Chimpanzee (100%) Gorilla, Orangutan, Gibbon (94%) Monkey (88%).
G protein-coupled receptors (GPCRs, or GPRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins.
Specifications
| GPR78 | |
| Polyclonal | |
| Unconjugated | |
| GPR78 | |
| G protein-coupled receptor 78; GPR78; G-protein coupled receptor 78; UNQ5925/PRO19818 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27201 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q96P69 | |
| GPR78 | |
| Synthetic 17 amino acid peptide from 3rd cytoplasmic domain of human GPR78. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction