Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR78 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582479
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582479 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582479 Supplier Invitrogen™ Supplier No. PA582479
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLAELVPFVT VNAQWGILSK CLTYSKAVAD PFTYSLLRRP FRQVLAGMVH RLLKRTPRPA STHDSSLDVA GMVHQLLKRT PRPASTHNGS VDTENDSCLQ QTH Highest antigen sequence indentity to the following orthologs: Mouse - 25%, Rat - 25%.

G protein-coupled receptors (GPCRs, or GPRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR78
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GPR78
Gene Accession No. Q96P69
Gene Alias G protein-coupled receptor 78; GPR78; G-protein coupled receptor 78; UNQ5925/PRO19818
Gene Symbols GPR78
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GPR78. Recombinant protein control fragment (Product #RP-89575).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27201
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.