Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR81 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120713
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120713 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120713 Supplier Invitrogen™ Supplier No. PA5120713
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HT-29, 293T, MCF7, Mouse brain, Rat liver. Immunogen sequence: SACDPSVHGA LHITLSFTYM NSMLDPLVYY FSSPSFPKFY NKLKICSLKP KQPGHSKTQR PEEMPISNLG RRSCISVANS FQSQSDGQWD PHIVEWH.

The G protein-coupled receptor GPCR GPR81 has been reported primarily in adipose and pituitary. ESTs have been isolated from human bladder cancer and colon cancer libraries. It has not been detected in frontal, temporal and occipital lobes of the cortex, basal forebrain, caudate nucleus, nucleus accumbens, and hippocampus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR81
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.96 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene HCAR1
Gene Accession No. Q8C131, Q9BXC0
Gene Alias FKSG80; G protein-coupled receptor 104; G protein-coupled receptor 81; GPR104; GPR81; G-protein coupled receptor 104; G-protein coupled receptor 81; HCA1; HCAR1; hydrocarboxylic acid receptor 1; hydroxycarboxylic acid receptor 1; hydroxy-carboxylic acid receptor 1; LACR1; lactate receptor 1; TAGPCR; TA-GPCR; T-cell activation G protein-coupled receptor
Gene Symbols HCAR1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 250-346 of human HCAR1 (NP_115943.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 243270, 27198, 369158
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.