Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR81 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA528734
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528734 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528734 Supplier Thermo Scientific Supplier No. PA528734

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, HeLa, Molt-4, Raji. Predicted reactivity: Rhesus Monkey (100%), Chimpanzee (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The G protein-coupled receptor GPCR GPR81 has been reported primarily in adipose and pituitary. ESTs have been isolated from human bladder cancer and colon cancer libraries. It has not been detected in frontal, temporal and occipital lobes of the cortex, basal forebrain, caudate nucleus, nucleus accumbens, and hippocampus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR81
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene HCAR1
Gene Accession No. Q9BXC0
Gene Alias FKSG80; G protein-coupled receptor 104; G protein-coupled receptor 81; GPR104; GPR81; G-protein coupled receptor 104; G-protein coupled receptor 81; HCA1; HCAR1; hydrocarboxylic acid receptor 1; hydroxycarboxylic acid receptor 1; hydroxy-carboxylic acid receptor 1; LACR1; lactate receptor 1; TAGPCR; TA-GPCR; T-cell activation G protein-coupled receptor
Gene Symbols HCAR1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a region within amino acids 1 and 45 of Human FKSG80
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27198
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.