Learn More
GPR83 Mouse anti-Mouse, Clone: OTI6C6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF811749

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
GPR83, a 423 A.A protein, is an orphan member of GPCR protein family and is closely related to tachykinin and NPY receptors. It is a human homolog of mouse GIR protein and is exclusively expressed and selectively up-regulated in brain. It is a cell surface receptor protein consisting of a putative seven-transmembrane-domain. Reports suggest GPR83 might be involved in the regulation of immune tolerance mechanism by inducing peripheral generation of Foxp3+ Treg cells.Specifications
GPR83 | |
Monoclonal | |
Unconjugated | |
Gpr83 | |
GIR, GPR72 | |
Mouse | |
Affinity Chromatography | |
RUO | |
14608 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI6C6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P30731 | |
Gpr83 | |
Synthetic peptide | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.