Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR83 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114742
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114742 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114742 Supplier Invitrogen™ Supplier No. PA5114742
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GPR83/GPR72.

GPR83, a 423 A.A protein, is an orphan member of GPCR protein family and is closely related to tachykinin and NPY receptors. It is a human homolog of mouse GIR protein and is exclusively expressed and selectively up-regulated in brain. It is a cell surface receptor protein consisting of a putative seven-transmembrane-domain. Reports suggest GPR83 might be involved in the regulation of immune tolerance mechanism by inducing peripheral generation of Foxp3+ Treg cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR83
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene GPR83
Gene Accession No. P30731, Q9NYM4
Gene Alias AW045253; G protein-coupled receptor 72; G protein-coupled receptor 83; Gir; glucocorticoid induced receptor; glucocorticoid-induced receptor; GPR72; Gpr83; G-protein coupled receptor 72; Hcepto9; Jp05; KIAA1540; Probable G-protein coupled receptor 83; RP105; RP39; RP82
Gene Symbols GPR83
Host Species Rabbit
Immunogen A synthesized peptide derived from human GPR83(Accession Q9NYM4), corresponding to amino acid residues S39-N89.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10888, 140595, 14608
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.