Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR84 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA3042
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA3042 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA3042 Supplier Invitrogen™ Supplier No. PA3042
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

IHC(P) analysis shows positive staining of GPR84 in human cerebral cortex and germinal center of human lymph node. WB analysis shows GPR84 in glyco-protein enriched fractions from GPR84 or GPR88 overexpressed 293 cells. The GPR84 antiserum showed specific detection of GPR84 over other tested GPRs. GPR84 exists in different glycosylated forms.

G protein-coupled receptors (GPCRs, or GPRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins. GPCR EX33 is a member of this family (subfamily Orphan A).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR84
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 0.05% sodium azide
Gene GPR84
Gene Accession No. Q9NQS5
Gene Alias EX33; G protein-coupled receptor 84; GPCR4; GPR84; G-protein coupled receptor 84; inflammation-related G protein-coupled receptor EX33; inflammation-related G-protein coupled receptor EX33; orphan G protein-coupled receptor 84
Gene Symbols GPR84
Host Species Rabbit
Immunogen KLH conjugated COOH-terminal amino acids of 377-396 (FRQAYGSILKRGPRSFHRLH).
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53831
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.