Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR84 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA130069
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA130069 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA130069 Supplier Invitrogen™ Supplier No. PA130069
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-30069 detects GPCR EX33 from human samples.

G protein-coupled receptors (GPCRs, or GPRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins. GPCR EX33 is a member of this family (subfamily Orphan A).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR84
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene GPR84
Gene Accession No. Q9NQS5
Gene Alias EX33; G protein-coupled receptor 84; GPCR4; GPR84; G-protein coupled receptor 84; inflammation-related G protein-coupled receptor EX33; inflammation-related G-protein coupled receptor EX33; orphan G protein-coupled receptor 84
Gene Symbols GPR84
Host Species Rabbit
Immunogen Synthetic 19 amino acid peptide from 2nd extracellular domain of human GPR84.
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100062469, 53831
Target Species Horse, Human, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.