Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR85 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533776
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533776 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533776 Supplier Invitrogen™ Supplier No. PA533776
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Orangutan, Mouse, Rat, Turkey, Chicken, Lizard, Xenopus, Seabass, Stickleback, Pufferfish, Zebrafish (100%).

GPR85, a member of the SREB (Super-Conserved Receptor Expressed in Brain) family of GPCRs, is an Orphan-A GPCR with an unknown ligand. The rat counterpart of SREB2 is involved in the development of the cerebral cortex. GPR85 has been reported to be expressed in several areas of human and rat CNS (Matsumoto et al., 2000), as well as in genital organs. ESTs have been isolated from B-cell/lung/testis, brain, testis, and embryo libraries, as well as many mouse tissue libraries.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR85
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.85 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene GPR85
Gene Accession No. P60893, P60894, P60895, Q9I919
Gene Alias 2900026B03Rik; FLJ50993; FLJ51159; FLJ99433; G protein-coupled receptor 85; Gpr85; PKrCx1; probable G-protein coupled receptor 85; Rgpr85; seven transmembrane helix receptor; SREB; SREB2; Srep2; Super conserved receptor expressed in brain 2
Gene Symbols GPR85
Host Species Rabbit
Immunogen Synthetic 20 amino acid peptide from 2nd extracellular domain of human GPR85.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 417775, 54329, 64020, 64450, 793299
Target Species Human, Mouse, Rat, Chicken, Frog, Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.