Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR85 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983707
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3707 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3707 Supplier Bioss Supplier No. BS15364R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GPR85, a member of the SREB (Super-Conserved Receptor Expressed in Brain) family of GPCRs, is an Orphan-A GPCR with an unknown ligand. The rat counterpart of SREB2 is involved in the development of the cerebral cortex. GPR85 has been reported to be expressed in several areas of human and rat CNS (Matsumoto et al., 2000), as well as in genital organs. ESTs have been isolated from B-cell/lung/testis, brain, testis, and embryo libraries, as well as many mouse tissue libraries.

Specifications

Antigen GPR85
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GPR85
Gene Accession No. P60893
Gene Alias 2900026B03Rik; BOS_4247; FLJ50993; FLJ51159; FLJ99433; G protein-coupled receptor 85; G protein-coupled receptor 85-like; GPR85; LOC100357476; PKrCx1; Probable G-protein coupled receptor 85; Rgpr85; seven transmembrane helix receptor; SREB; Sreb2; Srep2; Super conserved receptor expressed in brain 2
Gene Symbols GPR85
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GPR85.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54329
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.