Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR87 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP25686325UL
Description
GPR87 Polyclonal specifically detects GPR87 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
GPR87 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
G protein-coupled receptor 87, G protein-coupled receptor 95, GPR95, G-protein coupled receptor 87, G-protein coupled receptor 95, KPG_002, MGC131898, orphan GPCR 87 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
IgG |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
GPR87 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:CRSFSRRLFKKSNIRTRSESIRSLQSVRRSEVR | |
25 μL | |
GPCR | |
53836 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction