Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPR87 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533781
Description
Percent identity with other species by BLAST analysis: Human, Gibbon, Monkey, Marmoset, Mouse, Rat, Bovine, Dog, Bat, Hamster, Panda, Horse, Rabbit, Pig (100%).
G protein-coupled receptors play a role in cell communication. They are characterized by an extracellular N terminus, 7 transmembrane regions, and an intracellular C terminus. GPR87 has been reported to be expressed in human prostate and rat stomach and fetal tissues. ESTs have been isolated from B-cell/lung/testis, breast, heart, jaw, prostate, and testis libraries.
Specifications
| GPR87 | |
| Polyclonal | |
| Unconjugated | |
| GPR87 | |
| FKSG78; G protein-coupled receptor 87; G protein-coupled receptor 95; GPR87; GPR95; G-protein coupled receptor 87; G-protein coupled receptor 95; KPG_002; MGC131898; orphan GPCR 87 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100050286, 100344243, 100524399, 100770345, 310443, 485718, 53836, 785030, 84111 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q99MT7, Q9BY21 | |
| GPR87 | |
| Synthetic 19 amino acid peptide from C-terminus of human GPR87. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Hamster, Pig, Horse, Bat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction