Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPR87 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA120460
Description
PA1-20460 detects G Protein-Coupled Receptor GPR87 in human samples.
G protein-coupled receptors play a role in cell communication. They are characterized by an extracellular N terminus, 7 transmembrane regions, and an intracellular C terminus. GPR87 has been reported to be expressed in human prostate and rat stomach and fetal tissues. ESTs have been isolated from B-cell/lung/testis, breast, heart, jaw, prostate, and testis libraries.
Specifications
GPR87 | |
Polyclonal | |
Unconjugated | |
GPR87 | |
FKSG78; G protein-coupled receptor 87; G protein-coupled receptor 95; GPR87; GPR95; G-protein coupled receptor 87; G-protein coupled receptor 95; KPG_002; MGC131898; orphan GPCR 87 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
53836 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q9BY21 | |
GPR87 | |
Synthetic peptide corresponding to the N-terminal extracellular domain of human GPR87. | |
25 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction